Anti-ZNF148 Mouse Polyclonal Antibody
Catalog # ABNOH00007707-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 148
- Antigen symbol:ZNF148
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7707
- Antigen synonyms:HT-BETA|pHZ-52|ZFP148|ZBP-89|BERF-1|BFCOL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ZNF148
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human