Anti-TCF7 Mouse Monoclonal Antibody [clone: 1D2]
Catalog # ABNOH00006932-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor 7
- Antigen symbol:TCF7
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6932
- Antigen synonyms:T-cell specific|TCF7|TCF-1|HMG-box|TCF1|Transcription factor 7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TCF7
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human