Anti-CAPN15 Mouse Monoclonal Antibody [clone: 2H7]
Catalog # ABNOH00006650-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calpain 15
- Antigen symbol:CAPN15
- Clonality:Monoclonal
- Clone:2H7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:6650
- Antigen synonyms:SOLH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CAPN15
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human