Order Entry
Germany
ContactUsLinkComponent
Anti-SLC20A2 Mouse Monoclonal Antibody [clone: 4B1]
Anti-SLC20A2 Mouse Monoclonal Antibody [clone: 4B1]
Catalog # ABNOH00006575-M04
Supplier:  Abnova
undefined
Anti-SLC20A2 Mouse Monoclonal Antibody [clone: 4B1]
Catalog # ABNOH00006575-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 20 Member A2
  • Antigen symbol:
    Solute carrier family 20, member 2
  • Clonality:
    Monoclonal
  • Clone:
    4B1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6575
  • Antigen synonyms:
    PIT2|MLVAR|IBGC3|GLVR-2|PIT-2|IBGC1|GLVR2|RAM1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SLC20A2
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human