Order Entry
Germany
ContactUsLinkComponent
Anti-SRSF3 Mouse Monoclonal Antibody [clone: 2D2]
Anti-SRSF3 Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ABNOH00006428-M08
Supplier:  Abnova
undefined
Anti-SRSF3 Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ABNOH00006428-M08
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serine/arginine-rich Splicing Factor 3
  • Antigen symbol:
    SRSF3
  • Clonality:
    Monoclonal
  • Clone:
    2D2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6428
  • Antigen synonyms:
    SFRS3|Serine/arginine-rich splicing factor 3|Pre-mRNA-splicing factor SRP20|Splicing factor arginine/serine-rich 3|SRP20
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SRSF3
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat