Anti-RAPSN Mouse Polyclonal Antibody
ABNOH00005913-A01
: Abnova
- Antibody type:Primary
- Antigen name:Receptor-Associated Protein Of The Synapse
- Antigen symbol:RAPSN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5913
- Antigen synonyms:CMS1D|CMS1E|MGC3597|RAPSN|43 kda postsynaptic protein|43 kD receptor-associated protein of the synapse|RNF205|receptor-associated protein of the synapse|43kD|rapsyn|acetylcholine receptor-associated 43 kda protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: RAPSN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human