Order Entry
Germany
ContactUsLinkComponent
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 1G4]
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 1G4]
Catalog # ABNOH00005562-M01
Supplier:  Abnova
undefined
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 1G4]
Catalog # ABNOH00005562-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 1
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Monoclonal
  • Clone:
    1G4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5562
  • Antigen synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human