Anti-PIM1 Mouse Monoclonal Antibody [clone: 1E2]
Catalog # ABNOH00005292-M11
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pim-1 proto-oncogene, serine/threonine kinase
- Antigen symbol:PIM1
- Clonality:Monoclonal
- Clone:1E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:5292
- Antigen synonyms:PIM|Proviral Integration Site 1|PIM-1|PIM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PIM1
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human