Anti-NME3 Mouse Monoclonal Antibody [clone: 3G10]
Catalog # ABNOH00004832-M13
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nucleoside diphosphate kinase C
- Antigen symbol:NME3
- Clonality:Monoclonal
- Clone:3G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4832
- Antigen synonyms:NDK 3|NDP kinase 3|NDPKC|Nucleoside diphosphate kinase 3|NME3|DR-nm23|nm23-H3|Nucleoside diphosphate kinase C
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NME3
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human