Anti-CITED1 Mouse Monoclonal Antibody [clone: 6G8]
ABNOH00004435-M01
: Abnova
- Antibody type:Primary
- Antigen name:Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 1
- Antigen symbol:CITED1
- Clonality:Monoclonal
- Clone:6G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:4435
- Antigen synonyms:CITED1|Cbp/p300-interacting transactivator 1|Melanocyte-specific protein 1|MSG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CITED1
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat