Order Entry
Germany
ContactUsLinkComponent
Anti-MECP2 Mouse Monoclonal Antibody [clone: 4B6]
Anti-MECP2 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # ABNOH00004204-M01
Supplier:  Abnova
undefined
Anti-MECP2 Mouse Monoclonal Antibody [clone: 4B6]
Catalog # ABNOH00004204-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Methyl CpG binding protein 2
  • Antigen symbol:
    MECP2
  • Clonality:
    Monoclonal
  • Clone:
    4B6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4204
  • Antigen synonyms:
    RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MECP2
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat