Order Entry
Germany
ContactUsLinkComponent
 
Anti-HOXD8 Mouse Monoclonal Antibody [clone: 5E11]
  ABNOH00003234-M03
 :  Abnova
undefined
Anti-HOXD8 Mouse Monoclonal Antibody [clone: 5E11]
  ABNOH00003234-M03
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    homeobox D8
  • Antigen symbol:
    HOXD8
  • Clonality:
    Monoclonal
  • Clone:
    5E11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3234
  • Antigen synonyms:
    HOX5.4|HOX4E|HOX4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: HOXD8
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human