Anti-DOCK1 Mouse Monoclonal Antibody [clone: 3D10]
ABNOH00001793-M02
: Abnova
- Antibody type:Primary
- Antigen name:Dedicator Of Cytokinesis 1
- Antigen symbol:DOCK1
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:1793
- Antigen synonyms:DOCK180|ced5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DOCK1
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human