Anti-DDX1 Mouse Polyclonal Antibody
ABNOH00001653-A01
: Abnova
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box helicase 1
- Antigen symbol:DDX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1653
- Antigen synonyms:UKVH5d|DBP-RB|DEAD box protein retinoblastoma|ATP-dependent RNA helicase DDX1|DEAD box polypeptide 1|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1|DEAD box-1|DEAD box protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: DDX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human