Anti-RUNX2 Mouse Monoclonal Antibody [clone: 1D2]
ABNOH00000860-M05
: Abnova
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 2
- Antigen symbol:RUNX2
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:860
- Antigen synonyms:Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RUNX2
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human