Anti-ASCL1 Rabbit Polyclonal Antibody
ABNOH00000429-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Achaete-scute Homolog 1
- Antigen symbol:ASCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:429
- Antigen synonyms:ASH-1|ASH1|Mash1|BHLHA46|HASH1|Mash-1|hASH1|bHLHa46|Class A basic helix-loop-helix protein 46|Achaete-scute homolog 1
- Storage buffer:In 1x PBS, pH 7,4
- Sequence:MESSAKMESGGAGQQPQPQPQQPFLPPAACFFATAAAAAAAAAAAAAQSAQQQQQQQQQQQQAPQLRPAADGQPSGGGHKSAPKQVKRQRSSSPELMRCKRRLNFSGFGYSLPQQQPAAVARRNERERNRVKLVNLGFATLREHVPNGAANKKMSKVETLRSAVEYIRALQQLLDEHDAVSAAFQAGVLSPTISPNYSNDLNSMAGSPVSSYSSDEGSYDPLSPEEQELLDFTNWF
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:ASCL1 (NP_004307.2, 1 a.a. ~ 236 a.a) full-length human protein.
- Size:100 µg
- Pk:100 µg
Rabbit polyclonal antibody raised against a full-length human ASCL1 protein.
Recommended Dilutions: Western Blot (1 µg/ml)
Type: Primary
Antigen: ASCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human