Anti-RAB30 Mouse Monoclonal Antibody [clone: OTI3B1]
Catalog # ORIGTA505347
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:RAB30, member RAS oncogene family
- Antigen symbol:RAB30
- Clonality:Monoclonal
- Clone:OTI3B1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:22.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: RAB30 (RAB30, member RAS oncogene family)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human