Anti-NANP Mouse Monoclonal Antibody [clone: OTI4D5]
Catalog # ORIGTA504547
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:N-acetylneuraminic acid phosphatase
- Antigen symbol:NANP
- Clonality:Monoclonal
- Clone:OTI4D5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:27.6 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: NANP (N-acetylneuraminic acid phosphatase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human