Anti-C1S Mouse Monoclonal Antibody [clone: OTI4D9]
Catalog # ORIGTA504079
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:complement component 1, s subcomponent
- Antigen symbol:C1S
- Clonality:Monoclonal
- Clone:OTI4D9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:74.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100
Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: