Anti-LMAN1 Mouse Monoclonal Antibody [clone: OTI1A8]
Catalog # ORIGTA502137
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:lectin, mannose-binding, 1
- Antigen symbol:LMAN1
- Clonality:Monoclonal
- Clone:OTI1A8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:54.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LMAN1 (lectin, mannose-binding, 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: