Anti-VSIG2 Mouse Monoclonal Antibody [clone: OTI1H2]
Catalog # ORIGTA502124
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:V-set and immunoglobulin domain containing 2
- Antigen symbol:VSIG2
- Clonality:Monoclonal
- Clone:OTI1H2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:34.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100
Type: Primary
Antigen: VSIG2 (V-set and immunoglobulin domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human