Anti-EXD1 Mouse Monoclonal Antibody [clone: OTI4D7]
Catalog # ORIGTA502100
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Exonuclease 3'-5' domain containing 1
- Antigen symbol:EXD1
- Clonality:Monoclonal
- Clone:OTI4D7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:58.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: EXD1 (exonuclease 3'-5' domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human