Anti-PIM2 Mouse Monoclonal Antibody [clone: OTI10A12]
ORIGTA501064
: OriGene
- Antibody type:Primary
- Antigen name:Pim-2 oncogene
- Antigen symbol:PIM2
- Clonality:Monoclonal
- Clone:OTI10A12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:34.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100
Type: Primary
Antigen: PIM2 (pim-2 oncogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human