Anti-BRAF Mouse monoclonal antibody [clone: OTI1E2]
Catalog # ORIGTA500845
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:v-raf murine sarcoma viral oncogene homolog B
- Antigen symbol:BRAF
- Clonality:Monoclonal
- Clone:OTI1E2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:84.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human