Order Entry
Germany
ContactUsLinkComponent
Anti-MMP2 Mouse Monoclonal Antibody [clone: 2C1]
Anti-MMP2 Mouse Monoclonal Antibody [clone: 2C1]
Catalog # ORIGTA309752
Supplier:  OriGene
Anti-MMP2 Mouse Monoclonal Antibody [clone: 2C1]
Catalog # ORIGTA309752
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    2C1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS and 0,05% Sodium Azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 μL
  • Pk:
    0,1 ml

Specifications

About this item

Recommended Dilutions:

Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat