Anti-SIRT1 Goat Polyclonal Antibody
Catalog # ORIGTA303141
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 1
- Antigen symbol:SIRT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-EITEKPPRTQKE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml
Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human