Specifications
- Antibody type:Primary
- Antigen name:Fc epsilon Receptor I alpha (FceR1)
- Clonality:Monoclonal
- Clone:MAR-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Armenian hamster
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:none
- Gene ID:22052205
- Antigen synonyms:high affinity IgE receptor
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.
- Purified
The MAR-1 antibody is widely used both in flow cytometry and for depletion of cells in vitro / in vivo. Please choose the appropriate format for each application.
Type: Primary
Antigen: FCER1 (Fc fragment of IgE, high affinity I, receptor for)
Clonality: monoclonal
Clone: 42064
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse