Order Entry
Germany
ContactUsLinkComponent
Anti-CTLA4 Mouse monoclonal antibody [clone: BNI3]
Anti-CTLA4 Mouse monoclonal antibody [clone: BNI3]
Catalog # 70-1529-U100
Supplier:  Tonbo Biosciences
undefined
Anti-CTLA4 Mouse monoclonal antibody [clone: BNI3]
Catalog # 70-1529-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytotoxic T-lymphocyte-Associated Protein 4
  • Antigen symbol:
    CTLA4
  • Clonality:
    Monoclonal
  • Clone:
    BNI3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Macaque,
    Human
  • Cross adsorption:
    none
  • Gene ID:
    1493
  • Antigen synonyms:
    IDDM12|GRD4|GSE|CD|CTLA-4|ALPS5|cytotoxic T-lymphocyte associated protein 4|CD152|CELIAC3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The BNI3 antibody is specific for human CD152, commonly known as CTLA-4, a 33-37 kDa protein expressed as a homodimer on the surface of activated T and B cells, and on thymocytes. CTLA-4 is structurally similar, yet functionally disparate, to the T cell co-stimulatory molecule CD28. Both CTLA-4 and CD28 interact with the co-stimulatory molecules CD80 (B7-1) and CD86 (B7-2) on antigen-presenting cells, with CTLA-4 displaying a higher avidity than CD28. While CD28 typically delivers a potent co-stimulatory signal in support of T cell activation, CTLA-4 appears to act as a negative regulator of T cell activation and may contribute to the suppressor function of Treg cells.

CTLA-4 proteins may be initially sequestered within Golgi vesicles, from which they can be transferred to and from the cell surface, a mechanism by which Treg cells can selectively impart suppressive functions. The BNI3 antibody may be used for flow cytometric analysis of intracellular or surface CTLA-4 expression, and is also widely used for neutralization of CTLA-4 when expressed at the cell surface. The BNI3 antibody is reported to be cross-reactive with Baboon, Cynomolgus and Rhesus CTLA-4. Please choose the appropriate format for each application.

Type: Primary
Antigen: CTLA4 (cytotoxic T-lymphocyte-associated protein 4)
Clonality: monoclonal
Clone: BNI3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: