Order Entry
Germany
ContactUsLinkComponent
Anti-IL7R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: A7R34]
Anti-IL7R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: A7R34]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IL7R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: A7R34]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-1271-U100
  • 50-1271-U025
  • Antigen symbol:
  • IL17RA
  • IL17RA
  • Antigen name:
  • Interleukin 17 Receptor A
  • Interleukin 17 Receptor A
  • Antigen synonyms:
  • IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
  • IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • A7R34
  • A7R34
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16172
  • 16172
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow cytometry
  • Flow cytometry

Specifications

Related Information