About this item
The Hit3a antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing cell proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.
The Hit3a antibody is a widely used phenotypic marker for human T cells. In addition, binding/cross-linking of Hit3a antibody to CD3e can induce cell activation. The antibody has also been demonstrated to be cross-reactive with Chimpanzee CD3. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone: Hit3a
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Specifications
- Catalog No:
- 50-0039-T025
- 50-0039-T100
- Antigen symbol:
- CD3E
- CD3E
- Antigen name:
- CD3 Molecule Epsilon
- CD3 Molecule Epsilon
- Antigen synonyms:
- CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- Hit3a
- Hit3a
- Reactivity:
- Chimpanzee, Human
- Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 916
- 916
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Tested applications:
- Flow Cytometry
- Flow Cytometry