Order Entry
Germany
ContactUsLinkComponent
Anti-CD3E Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Anti-CD3E Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD3E Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 40-0031-U500
  • 40-0031-M001
  • 40-0031-U100
  • Antigen symbol:
  • CD3E
  • CD3E
  • CD3E
  • Antigen name:
  • CD3 Molecule Epsilon
  • CD3 Molecule Epsilon
  • CD3 Molecule Epsilon
  • Antigen synonyms:
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 145-2C11
  • 145-2C11
  • 145-2C11
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Armenian hamster
  • Armenian hamster
  • Armenian hamster
  • Gene ID:
  • 12501
  • 12501
  • 12501
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice
  • Tested applications:
  • Functional Assays, Immunofluorescence Microscopy
  • Functional Assays, Immunofluorescence Microscopy
  • Functional Assays, Immunofluorescence Microscopy

Specifications

Related Information