About this item
The GARP5 antibody reacts with human GARP, also known as LRRC32 or Garpin, an 80 kDa transmembrane protein which acts as a receptor for the latent form of TGF-beta 1 (pro-TGF-beta), preventing its secretion. Specifically, GARP is reported to associate with the pro-domain of TGF-beta 1, known as latency-associated peptide (LAP), which must be cleaved to release the biologically active cytokine. The putative role of GARP in sequestering the TGF-beta is important for regulating its activity, as TGF-beta signaling is involved in development and function of Th17, Treg and many other immune cell types. GARP expression has been shown on platelets and at high levels on Foxp3+ Treg cells, where it is proposed to be a phenotypic identifier for activated human Treg cells.
The GARP5 antibody may be used for analysis of human GARP by flow cytometry, immunoprecipitation and Western blotting. Please choose the appropriate format for each application.
Type: Primary
Antigen: LRRC32 (leucine rich repeat containing 32)
Clonality: monoclonal
Clone: GARP5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 35-9882-T100
- 35-9882-T025
- Antigen symbol:
- LRRC32
- LRRC32
- Antigen name:
- Leucine Rich Repeat Containing 32
- Leucine Rich Repeat Containing 32
- Antigen synonyms:
- GARP|D11S833E
- GARP|D11S833E
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- GARP5
- GARP5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2615
- 2615
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Shipping temperature:
- Ice
- Ice
- Tested applications:
- Flow Cytometry
- Flow Cytometry