Order Entry
Germany
ContactUsLinkComponent
Anti-Ly-6G Rat Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: 1A8]
Anti-Ly-6G Rat Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: 1A8]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-Ly-6G Rat Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: 1A8]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-1276-U025
  • 25-1276-U100
  • Antigen symbol:
  • LY6G
  • LY6G
  • Antigen name:
  • Lymphocyte Antigen 6 Complex Locus G
  • Lymphocyte Antigen 6 Complex Locus G
  • Antigen synonyms:
  • Gr1|Gr-1|Ly-6G
  • Gr1|Gr-1|Ly-6G
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • 1A8
  • 1A8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 546644
  • 546644
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information