Order Entry
Germany
ContactUsLinkComponent
Anti-CD3D Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: UCHT1]
Anti-CD3D Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: UCHT1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD3D Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: UCHT1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-0038-T025
  • 25-0038-T100
  • Antigen symbol:
  • CD3D
  • CD3D
  • Antigen name:
  • CD3 Molecule Delta
  • CD3 Molecule Delta
  • Antigen synonyms:
  • CD3-DELTA|IMD19|T3D
  • CD3-DELTA|IMD19|T3D
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • UCHT1
  • UCHT1
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 915
  • 915
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information