Order Entry
Germany
ContactUsLinkComponent
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-1172-U025
  • 20-1172-U100
  • Antigen symbol:
  • KIT
  • KIT
  • Antigen name:
  • Tyrosine-protein kinase Kit
  • Tyrosine-protein kinase Kit
  • Antigen synonyms:
  • Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
  • Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • ACK2
  • ACK2
  • Reactivity:
  • Rat, Mouse
  • Rat, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16590
  • 16590
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information