About this item
The PC61.5 antibody is specific for mouse CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Expression of CD25 varies during developmental stages of T and B cells, is induced on activated mature T and B cells, and is present on subsets of dendritic cells. CD25 signaling as part of the IL-2 receptor complex triggers T cell activation and proliferation, as well as modulating the differentiation and function of Th17 cells, T regulatory (Treg) cells, and dendritic cells.
The PC61.5 antibody is used as a marker for T cells, B cells and dendritic cell subsets. Expression of CD25, CD4 and the transcription factor Foxp3 is regarded as a phenotypic signature for Treg cells. As such, this antibody is widely used to distinguish Treg cells from naïve or conventional T cells which are CD25-. This clone has also been reported for depletion of Treg cells in vivo (use format suitable for functional assays).
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: PC61.5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1 Lambda
Reactivity: Mouse
Specifications
- Catalog No:
- 20-0251-U025
- 20-0251-U100
- Antigen symbol:
- IL2RA
- IL2RA
- Antigen name:
- Interleukin 2 Receptor, Alpha
- Interleukin 2 Receptor Alpha
- Antigen synonyms:
- TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- PC61.5
- PC61.5
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16184
- 16184
- Isotype:
- IgG1 lambda
- IgG1 lambda
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice