Order Entry
Germany
ContactUsLinkComponent
 
Anti-ALDH3B1 Rabbit Polyclonal Antibody
  ABGEAP8706C
 :  Abgent
undefined
Anti-ALDH3B1 Rabbit Polyclonal Antibody
  ABGEAP8706C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Aldehyde Dehydrogenase 3 Family, member B1
  • Antigen symbol:
    ALDH3B1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Gene ID:
    5439
  • Antigen synonyms:
    RPB11|hRPB14|RPB11m|RPB11A|POLR2J1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    CRFRVAIKDKQEGIE
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This POLR2J antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 89-117 amino acids from the C-terminal region of human POLR2J.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ALDH3B1 (aldehyde dehydrogenase 3 family, member B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human