Order Entry
Germany
ContactUsLinkComponent
Anti-IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon) Rabbit Polyclonal Antibody [clone: 421CT8.1.1]
Anti-IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon) Rabbit Polyclonal Antibody [clone: 421CT8.1.1]
Catalog # ABGEAP8138B
Supplier:  Abgent
undefined
Anti-IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon) Rabbit Polyclonal Antibody [clone: 421CT8.1.1]
Catalog # ABGEAP8138B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Alkaline Phosphatase, Placental
  • Antigen symbol:
    IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon)
  • Clonality:
    Polyclonal
  • Clone:
    421CT8.1.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal (700–800 aa)
  • Cross adsorption:
    No
  • Form:
    Ascites
  • Gene ID:
    250
  • Antigen synonyms:
    PALP|ALP|PLAP|PLAP-1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    VTAARILKGQKKDC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    This ALPP antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 56-83 amino acids from the N-terminal region of human ALPP.
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human