Anti-SGK3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7949A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Serum/glucocorticoid regulated kinase 3
- Antigen symbol:SGK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal (13–30 aa)
- Cross adsorption:No
- Gene ID:26397
- Antigen synonyms:Serine/threonine-protein kinase SGK3|Serum/glucocorticoid regulated kinase-like SGKL|serum/glucocorticoid regulated kinase 3
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:LDKFYRKVLEKNMKC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This Mouse Map2k3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 132-160 amino acids from the Central region of mouse Map2k3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SGK3 (serum/glucocorticoid regulated kinase family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human