Anti-CYP2F1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7883B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily F, Polypeptide 1
- Antigen symbol:CYP2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:CYP2F|C2F1|CYPIIF1|Cytochrome P450 2F1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CYP2F1 (cytochrome P450, family 2, subfamily F, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human