Anti-CAMK1G (CaMKI gamma) Rabbit Polyclonal Antibody
ABGEAP7253B
: Abgent
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent Protein Kinase I Gamma
- Antigen symbol:CAMK1G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (434–450 aa)
- Cross adsorption:No
- Antigen synonyms:VWS1|CLICKIII|CLICK3|dJ272L16.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CAMK1G (CaMKI gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human