Anti-Hsp 60 Rabbit Polyclonal Antibody
Catalog # ABGEAP6800A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 70 kDa protein 14
- Antigen symbol:HSPA14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:HSP70-like protein 1|HSP60|Heat shock protein HSP60|HSP70L1|HSP7E|Heat shock 70 kDa protein 14|HSPA14
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Hsp 60
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse