Order Entry
Germany
ContactUsLinkComponent
Anti-AMER1 Rabbit Polyclonal Antibody
Anti-AMER1 Rabbit Polyclonal Antibody
  ABGEAP6553C
 :  Abgent
undefined
Anti-AMER1 Rabbit Polyclonal Antibody
  ABGEAP6553C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 123, member B
  • Antigen symbol:
    FAM123B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    Islet/duodenum homeobox-1|IPF-1|Insulin promoter factor 1|Pancreas/duodenum homeobox protein 1|IPF1|Somatostatin-transactivating factor 1|Insulin upstream factor 1|IUF-1|PDX-1|GSF|STF-1|Glucose-sensitive factor|PDX1|IDX-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: AMER1 (APC membrane recruitment protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse