Anti-ARF1 Rabbit Polyclonal Antibody
ABGEAP5871C
: Abgent
- Antibody type:Primary
- Antigen name:ADP-Ribosylation Factor 1
- Antigen symbol:ARF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:ADP-ribosylation factor 1|ADP Ribosylation Factor 1|DJ133P16.1.|ARF 1|ARF1_HUMAN|ARF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ARF1 (ADP-ribosylation factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human