Order Entry
Germany
ContactUsLinkComponent
Anti-ENAM Rabbit Polyclonal Antibody
Anti-ENAM Rabbit Polyclonal Antibody
Catalog # ABGEAP5838B
Supplier:  Abgent
undefined
Anti-ENAM Rabbit Polyclonal Antibody
Catalog # ABGEAP5838B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Rapamycin-insensitive companion of mTOR
  • Antigen symbol:
    ENAM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    AVO3 homolog|AVO3|PIA|RICTOR|Rapamycin-insensitive companion of mTOR|KIAA1999|hAVO3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ENAM (enamelin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human