Anti-TGIF1 Rabbit Polyclonal Antibody
Catalog # ABGEAP14571A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:TGFB-induced factor homeobox 1
- Antigen symbol:TGIF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:7050
- Antigen synonyms:5'-TG-3'-interacting factor 1|Homeobox protein TGIF1|TGIF
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:43.013 kDa
- Sequence:PELGTKAGPRRPHRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This TGIF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 81-109 amino acids from the N-terminal region of human TGIF1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TGIF1 (TGFB-induced factor homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human