Anti-MED8 Rabbit Polyclonal Antibody
Catalog # ABGEAP12649C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 8
- Antigen symbol:MED8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:DGAT2L5|FLJ22644|MGC119184|MGAT2|MGC119183|monoacylglycerol O-acyltransferase 2|MOGAT2|MGC119185|acyl CoA:monoacylglycerol acyltransferase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MED8 (mediator complex subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human