Anti-EEF1A2 Rabbit Polyclonal Antibody
Catalog # ABGEAP12557B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation elongation factor 1 alpha 2
- Antigen symbol:EEF1A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:HS1|STN|EF1A|eukaryotic elongation factor 1 A-2|STNL|eEF1A-2|EF-1-alpha-2|EEF1AL|statin S1|elongation factor 1-alpha 2|elongation factor-1 alpha|statin-S1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: EEF1A2 (eukaryotic translation elongation factor 1 alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse