Anti-NIPAL4 Rabbit Polyclonal Antibody
Catalog # AVIVARP67698_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:NIPA-like domain containing 4
- Antigen symbol:NIPAL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:348938
- Antigen synonyms:ARCI6|ICHTHYIN|ICHYN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NIPAL4 (NIPA-like domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human