Anti-MDGA2 Rabbit Polyclonal Antibody
AVIVARP66393_P050
- Antibody type:Primary
- Antigen name:MAM domain containing glycosylphosphatidylinositol anchor 2
- Antigen symbol:MDGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:161357
- Antigen synonyms:c14_5286|MAMDC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MDGA2 (MAM domain containing glycosylphosphatidylinositol anchor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human